Calcium stearate NF

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: Stearic acid calcium salt

CA169-45KGBL CA169-2.5KG CA169-12KG CA169-500GM
89078-676EA 1395.25 USD
89078-676 95032-338 89078-674 95032-340
Calcium stearate NF
Calcium stearate

Calcium Stearate, NF has good anti-adherent and lubricant properties. 


All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP). 

Formula: C₃₆H₇₀CaO₄
MW: 607.03 g/mol
Melting Pt: 147…149 °C
Density: 1.065 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00036390
CAS Number: 1592-23-0
EINECS: 216-472-8
Merck Index: 13,01708
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=66064c431fc65dcdf98ff2bc1d0227f37de78108&vid=c1765b9467ae214d145f3824443319aa43d8bd1e&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR