Citral for synthesis

Supplier: SPECTRUM CHEMICAL MFG CORP
Warning

Synonyms: 3,7-Dimethylocta-2,6-dienal

C2233-500ML C2233-100ML C2233-200LTB
700003-670EA 242.85 USD
700003-670 700003-666 700003-668
Citral for synthesis
Citral

Citral, also known as 3,7-dimethyl-2,6-octadienal or lemonal, is used in certain perfumes for its citrus effect and is considered to have strong antimicrobial qualities.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: C₁₀H₁₆O
MW: 152.24 g/mol
Boiling Pt: 229
Melting Pt: < –10 °C
Density: 0.888
Flash Pt: 98 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00006997
CAS Number: 5392-40-5
EINECS: 226-394-6
Merck Index: 13,02346

Order Now

Specification Test Results

Assay 96% Min.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR