(S)-(+)-Carvone ≥96%

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: D-p-Mentha-1(6),8-dien-2-one, D(+)-Carvone, (S)-2-Methyl-5-(prop-1-en-2-yl)cyclohex-2-enone, (S)-(+)-2-Methyl-5-(1-methylvinyl)cyclohex-2-en-1-one

C1693-100GM C1693-25GM C1693-1KG
700003-984EA 272.7 USD
700003-984 700003-988 700003-986
(S)-(+)-Carvone ≥96%
(S)-(+)-Carvone

D-Carvone, also known as δ6:8(9)-p-menthadien-2-one, is found in several essential oils and part of a family of chemicals called terpenoids; it is used in the flavor and food industries.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: C₁₀H₁₄O
MW: 150.22 g/mol
Boiling Pt: 230 °C (1013 hPa)
Melting Pt: ? 15 °C
Density: 0.9584 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00062997
CAS Number: 2244-16-8
EINECS: 218-827-2

Order Now

Specification Test Results

Assay 96% Min.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR