trans-1,2-Diaminocyclohexane-N,N,N',N'-tetraacetic acid monohydrate ≥98%, Reagent Grade
Supplier: SPECTRUM CHEMICAL MFG CORP
|
Warning
|
Synonyms:
Titriplex® IV
C1466-12KGBL
C1466-100GM
C1466-25GM
C1466-500GM
700007-240EA
10779.35
USD
700007-240
700007-238
700007-242
700007-244
trans-1,2-Diaminocyclohexane-N,N,N',N'-tetraacetic acid monohydrate ≥98%, Reagent Grade
trans-1,2-Diaminocyclohexane-N,N,N',N'-tetraacetic acid monohydrate
(1,2-Cyclohexylenedinitrilo)tetraacetic Acid, Reagent is an alkyl-substituted amino acid that functions as a chelating agent. This ingredient's source is synthetic. It appears as a solid and is partially soluble in cold water. Its reagent grade means this is the highest quality commercially available for this chemical and that the American Chemical Society has not officially set any specifications for this material.
Formula:
C₁₄H₂₂N₂O₈·1H₂O MW: 364.35 g/mol Melting Pt: 213…216 °C Storage Temperature: Ambient |
MDL Number:
MFCD00066429 CAS Number: 125572-95-4 EINECS: 236-308-9 |
Specification Test Results
ASSAY (C14H22N2O8•H2O) | 98% Min. |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...