Vitamin B12 (cyanocobalamin) FCC
Supplier: SPECTRUM CHEMICAL MFG CORP
C1454-1KGBL
C1454-5GM
C1454-100GMB
C1454-1GM
C1454-25GM
C1454-500MG
95032-748EA
17556.3
USD
95032-748
95032-754
95032-744
95032-746
95032-750
95032-752
Vitamin B12 (cyanocobalamin) FCC
Vitamin B12 (cyanocobalamin)
Vitamin B12, FCC is used as a dietary supplement. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage, nutritional supplements and personal care applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Formula:
C₆₃H₈₈CoN₁₄O₁₄P MW: 1355.38 g/mol Melting Pt: 320 °C Storage Temperature: Ambient |
MDL Number:
MFCD00151092 CAS Number: 68-19-9 EINECS: 200-680-0 Merck Index: 12,10152 |
Specification Test Results
Assay | 96% Min. |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...