Chromium(III) oxide, green ≥98%, powder, purified

Supplier: Spectrum Chemical Mfg. Corp.

C1255-500GM C1255-125GM C1255-2.5KG C1255-12KG
700000-836EA 378.55 USD
700000-836 700000-828 700000-832 700000-830
Chromium(III) oxide, green ≥98%, powder, purified
Chromium(III) oxide

Chromium Oxide, Green, Powder, Purified is often used in industrial paints where a lot of wear is involved.


The Purified Grade describes chemicals of good quality where there are no official standards and is normally limited to inorganic chemicals.

Formula: Cr₂O₃
Boiling Pt: 4000 °C (1013 hPa)
Melting Pt: 2435 °C
Density: 5.22 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010949
CAS Number: 1308-38-9
EINECS: 215-160-9
Merck Index: 13,02247

Order Now

Specification Test Results

Assay 98% Min.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR