Cerium(IV) oxide ≥95%, Reagent Grade for synthesis

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: Ceric oxide, Cerium dioxide

C1202-100GM C1202-25KGBL
700002-094EA 250.3 USD
700002-094 700002-096
Cerium(IV) oxide ≥95%, Reagent Grade for synthesis
Cerium(IV) oxide

Ceric Oxide, Reagent, also known as cerium (IV) oxide or ceria, can be used as an oxidizing species in catalytic converters. The Reagent grade denotes that this chemical is the highest quality commercially available and that the American Chemical Society has not officially set any specifications for this material. Spectrum Chemical manufactured Reagent grade products meet the toughest regulatory standards for quality and purity.

Formula: CeO₂
MW: 172.11 g/mol
Boiling Pt: 3468
Melting Pt: 2000
Density: 7.13 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010933
CAS Number: 1306-38-3
EINECS: 215-150-4
Merck Index: 12,02035

Order Now

Specification Test Results

Assay 95% Min.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR