D-(+)-Camphor ≥98%

Supplier: Spectrum Chemical Mfg. Corp.
Warning

C1164-500GM C1164-100GM
700003-960EA 119.78 USD
700003-960 700003-958
D-(+)-Camphor ≥98%
D-(+)-Camphor

D-(+)-Camphor, also known as 2-Camphanone, is a terpinoid with a strong aromatic odor. This waxy, flammable, solid is used as an antimicrobial, a moth repellent and is readily absorbed through the skin producing a feeling of cooling similar to that of menthol. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.

Formula: C₁₀H₁₆O
Boiling Pt: 408 °C (1013 hPa)
Melting Pt: 174…175 °C
Density: 0.992 g/cm³ (20 °C)
Flash Pt: 66 °C
Storage Temperature: Ambient
MDL Number: MFCD00064149
CAS Number: 464-49-3
EINECS: 207-355-2
UN: 2717
ADR: 4.1,III
Merck Index: 13,01739

Order Now

Specification Test Results

Assay 98% Min.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR