Calcium phosphate tribasic, powder, Reagent Grade suitable for use as excipient

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: Hydroxylapatite, tri-Calcium phosphate, HAp, Calcium Hydroxide Phosphate, Calcium hydroxyphosphate, Tribasic calcium phosphate

C1155-12KG C1155-45KGBL C1155-125GM C1155-2.5KG C1155-500GM
700000-358EA 3080.9 USD
700000-358 700000-362 700000-356 700000-360 700000-364
Calcium phosphate tribasic, powder, Reagent Grade suitable for use as excipient
Calcium phosphate tribasic

Calcium Phosphate Tribasic, Powder, Reagent can also be referred to as Tricalcium phosphate and is abbreviated TCP. It is one of the main products of bone combustion but can also be obtained from inorganic sources such as mineral rock. It is used in baby powder as well as in powdered spices as an anti-caking agent. It is also used in gene transfection as well as for repairing bony defects and as a diet supplement. It has also historically been used in the production of porcelain. Its reagent grade means this is the highest quality commercially available for this chemical and that the American Chemical Society has not officially set any specifications for this material.

Formula: Ca₅HO₁₃P₃
MW: 502.306 g/mol
Density: 3.14 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010904
CAS Number: 12167-74-7
EINECS: 231-840-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR