Carboxymethyl cellulose sodium salt ≥99.5% FCC, low viscosity

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: Carboxymethyl cellulose, sodium salt, Cellulose gum, Cellulose, carboxymethyl ether, sodium salt, Sodium carboxymethyl cellulose (Viscosity 800mpa.s), Sodium carboxymethyl cellulose, CMC

C1103-500GM C1103-125GM C1103-2.5KG
95032-426EA 393.4 USD
95032-426 95032-422 95032-424
Carboxymethyl cellulose sodium salt ≥99.5% FCC, low viscosity
Carboxymethyl cellulose sodium salt

Cellulose Gum, Low Viscosity, FCC is used as a thickener and stabilizer in food products. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food Grade (FCC) chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Formula: [C₆H₇O₂(OH)x(OCH₂COONa)y]n
Melting Pt: 300 °C
Storage Temperature: Ambient
MDL Number: MFCD00081472
CAS Number: 9004-32-4
Merck Index: 12,01877

Order Now

Specification Test Results

Assay 99.5% Min.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR