Calcium citrate tetrahydrate ≥97.5%, powder USP
Supplier: SPECTRUM CHEMICAL MFG CORP
Synonyms:
tri-Calcium dicitrate tetrahydrate, Calcium citrate tribasic tetrahydrate
C1102-125LBB
C1102-2.5KG
C1102-500GM
C1102-12KG
700006-174EA
2917
USD
700006-174
95032-258
95032-260
700006-176
Calcium citrate tetrahydrate ≥97.5%, powder USP
Calcium citrate tetrahydrate
Form: Powder
All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Formula:
C₁₂H₁₀Ca₃O₁₄·4H₂O MW: 570.5 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00150786 CAS Number: 5785-44-4 EINECS: 212-391-7 |
Specification Test Results
Assay | 97.5% Min. |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...