Butyl-4-hydroxybenzoate ≥98% NF

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: Butyl 4-hydroxybenzoate, 4-Hydroxybenzoic acid butyl ester, Butyl parahydroxybenzoate, Butylparaben

BU115-12KG BU115-125GM BU115-500GM BU115-2.5KG BU115-50KGBL
89078-648EA 2911.15 USD
89078-648 95032-186 95032-190 95032-188 89078-650
Butyl-4-hydroxybenzoate ≥98% NF
Butyl-4-hydroxybenzoate

BSE/TSE statement (available upon request) Allergen statement (available upon request)


All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP). 

Formula: C₁₁H₁₄O₃
MW: 194.23 g/mol
Boiling Pt: 156…157 °C (3.5 mmHg)
Melting Pt: 73 °C
Density: 1.23 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00016478
CAS Number: 94-26-8
EINECS: 202-318-7
Merck Index: 13,01583

Order Now

Specification Test Results

Assay 98% Min.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR