Bromophenol blue ACS
Supplier: SPECTRUM CHEMICAL MFG CORP
Synonyms:
Bromphenol Blue Sultone Form, BPB, 3,3',5,5'-Tetrabromophenolsulphonphthalein, Tetrabromophenol-sulphonephthalein
Bromophenol Blue, Reagent, ACS is used as an acid-base indicator and a gel electrophoresis color marker. As an acid-base indicator its useful range lies between pH 3.0 and 4.6. At pH 3.0 it is blue and at pH 4.6 it changes to yellow. The reaction is reversible.
As an ACS grade quality reagent, its chemical specifications are the de facto standards for chemicals used in many high-purity applications and typically designate the highest quality chemical available for laboratory use. Spectrum Chemical manufactured Reagent ACS grade products meet the toughest regulatory standards for quality and purity.
Formula:
C₁₉H₁₀Br₄O₅S MW: 669.97 g/mol Melting Pt: 273 °C Density: 1 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00005875 CAS Number: 115-39-9 EINECS: 204-086-2 Merck Index: 13,01429 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...