tert-Butyl-4-methoxyphenol (mixture of isomers) ≥98.5% FCC suitable for use as excipient
Supplier: SPECTRUM CHEMICAL MFG CORP
Synonyms:
tert-Butyl-4-methoxyphenol, BHA, tert-Butyl-4-hydroxyanisole, Butylhydroxyanisole, 2(3)-tert-Butyl-4-methoxyphenol, Butylated hydroxyanisole
BH104-12KGBL
BH104-125GM
BH104-2.5KG
BH104-500GM
BH104-45KGBL
89078-620EA
3361.58
USD
89078-620
95032-072
95032-074
95032-076
89078-622
tert-Butyl-4-methoxyphenol (mixture of isomers) ≥98.5% FCC suitable for use as excipient
tert-Butyl-4-methoxyphenol (mixture of isomers)
BHA, FCC is an antioxidant and preservative used in food, food packaging and animal feed. Spectrum Chemical offers over 300 Food Grade (FCC) chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities
Formula:
C₁₁H₁₆O₂ Boiling Pt: 264 °C (1013 hPa) Melting Pt: 48…55 °C Density: 1.05 g/cm³ (20 °C) Flash Pt: 156 °C Storage Temperature: Ambient |
MDL Number:
MFCD01779059 CAS Number: 25013-16-5 EINECS: 246-563-8 Merck Index: 12,01582 |
Specification Test Results
Assay | 98.5% Min. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...