Benzyl benzoate ≥99% USP

Supplier: SPECTRUM CHEMICAL MFG CORP
Warning

Synonyms: Benzoic acid benzyl ester

BE159-4LTGL BE159-20LT BE159-20LTBL BE159-500MLG BE159-200LTB BE159-200LTB
95032-000EA 651.6 USD
95032-000 700005-564 89078-616 700005-566 700005-562 89078-618
Benzyl benzoate ≥99% USP
Benzyl benzoate

BSE/TSE statement (available upon request) Allergen statement (available upon request)


All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Formula: C₁₄H₁₂O₂
MW: 212.25 g/mol
Boiling Pt: 323 °C (1013 hPa)
Melting Pt: 17…20 °C
Density: 1.118 g/cm³ (20 °C)
Flash Pt: 158 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00003075
CAS Number: 120-51-4
EINECS: 204-402-9
UN: 3082
ADR: 9,III
Merck Index: 13,01129

Order Now

Specification Test Results

Assay 99% Min.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR