Benzyl alcohol ≥98% NF

Supplier: SPECTRUM CHEMICAL MFG CORP
Warning

Synonyms: Benzenemethanol, Phenylmethanol, Phenylcarbinol

BE158-4LTGL BE158-500MLG BE158-500MLG BE158-200LTB BE158-200LTB
95031-996EA 1059.05 USD
95031-996 700003-142 95031-998 700003-140 89078-614
Benzyl alcohol ≥98% NF
Benzyl alcohol

Benzyl Alcohol, NF is a synthetic ingredient used as solvent and preservative. 


All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP). 

Formula: C₆H₅CH₂OH
MW: 108.14 g/mol
Boiling Pt: 205 °C (1013 hPa)
Melting Pt: –15.3 °C
Density: 1.05 g/cm³ (20 °C)
Flash Pt: 101 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00004599
CAS Number: 100-51-6
EINECS: 202-859-9
Merck Index: 13,01126
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b0c85c958ac64f8b86c5836e49f2c4354b22886d&vid=633ad635461465914e782d5a22480c9ff8f81bf7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 98% Min.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR