1-Docosanol

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: Behenyl alcohol

B2859-100GM B2859-10GM
700002-742EA 211 USD
700002-742 700002-744
1-Docosanol
1-Docosanol

Behenyl Alcohol is a natural vegetable source saturated fatty alcohol used to regulate viscosity in formulations. It functions as a thickener and stabilizer and may be used as a co-emulsifier. May be used in water-in-oil emulsions, oil-in-water emulsions, and anhydrous formulations such as ointments, body butters and scrubs. It will produce elegant emulsions and impart a soft, velvety feel to the skin.

Formula: H₃C(CH₂)₂₀CH₂OH
MW: 326.61 g/mol
Boiling Pt: 376 °C (1013 hPa)
Melting Pt: 68…71 °C
Density: ~ 0.81 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002939
CAS Number: 661-19-8
EINECS: 211-546-6
Merck Index: 13,03433

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR