Benzyl salicylate ≥98%
Supplier: Spectrum Chemical Mfg. Corp.
Benzyl Salicylate, a salicylic acid benzyl ester, can be used as a component in perfumes and as a solvent for crystalline synthetic musks.
Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.
Formula:
C₁₄H₁₂O₃ Boiling Pt: 208 °C (1013 hPa) Melting Pt: 23.9 °C Density: 1.176 Storage Temperature: Ambient |
MDL Number:
MFCD00020034 CAS Number: 118-58-1 EINECS: 204-262-9 Merck Index: 13,01146 |
Specification Test Results
Assay | 98% Min. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...