Benzyl-4-hydroxybenzoate suitable for use as excipient
Supplier: SPECTRUM CHEMICAL MFG CORP
|
Warning
|
Synonyms:
Benzyl 4-hydroxybenzoate, Benzylparaben, 4-Hydroxybenzoic acid benzyl ester, Benzyl Parahydroxybenzoate
Benzylparaben is one of the parabens family. Parabens are esters formed by p-hydroxybenzoic acid and an alcohol. They are largely used as biocides in cosmetics and toiletries, medicaments, or food. They have synergistic power with other biocides.
Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.
Formula:
C₁₄H₁₂O₃ Storage Temperature: Ambient |
MDL Number:
MFCD00016471 CAS Number: 94-18-8 EINECS: 202-311-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...