Oxybenzone ≥97% USP

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: Eusolex® 4360, 2-Hydroxy-4-methoxybenzophenone, (2-Hydroxy-4-methoxyphenyl)(phenyl)methanone

B1261-12KGBL B1261-50KGBL B1261-100GM B1261-25GM B1261-500GM
700004-126EA 1906.25 USD
700004-126 700004-128 95034-812 95034-814 95034-816
Oxybenzone ≥97% USP
Oxybenzone

BSE/TSE statement (available upon request) Allergen statement (available upon request)


All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Formula: C₁₄H₁₂O₃
MW: 228.25 g/mol
Boiling Pt: 370 °C (1013 hPa)
Melting Pt: 65 °C
Density: 1.3397 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00008387
CAS Number: 131-57-7
EINECS: 205-031-5
Merck Index: 12,07088

Order Now

Specification Test Results

Assay 97% Min.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR