Benzyl nicotinate
Supplier: SPECTRUM CHEMICAL MFG CORP
|
Warning
|
Synonyms:
3-Picolinic acid benzyl ester, Pyridine-3-carboxylic acid benzyl ester, Benzyl 3-pyridinecarboxylate, 3-Pyridinecarboxylic acid benzyl ester, Nicotinic acid benzyl ester
B1212-250GM
B1212-50GM
700008-700EA
1331.7
USD
700008-700
700008-702
Benzyl nicotinate
Benzyl nicotinate
Benzyl Nicotinate is a reactant used in the synthesis of indolizine derivatives.
Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.
Formula:
C₁₃H₁₁NO₂ MW: 213.24 g/mol Boiling Pt: 179…180 °C (8 hPa) Melting Pt: 24 °C Density: 1.17 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00023584 CAS Number: 94-44-0 EINECS: 202-332-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...