1-Butanol ≥99.4% ACS
Supplier: Spectrum Chemical Mfg. Corp.
|
Danger
|
Butyl Alcohol, Reagent, ACS is more commonly known as butanol. It is also widely used as a solvent in textile and chemical fabrication processes. It can be used as a paint thinner and base for perfumes. As an ACS grade reagent, Spectrum Chemical manufactured compound is used as the quality standard against which other substances are grade and has met the toughest regulatory standards for quality and pureness
Formula:
H₃C(CH₂)₂CH₂OH Boiling Pt: 117.6 °C (1013 hPa) Melting Pt: –89 °C Density: 0.812 g/cm³ (20 °C) Flash Pt: 30 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002964 CAS Number: 71-36-3 EINECS: 200-751-6 UN: 1120 ADR: 3,III REACH: 01-2119484630-38 Merck Index: 13,01539 |
|
Specification Test Results
Assay | 99.4% Min. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...