Diethylene glycol monobutyl ether ≥99%

Supplier: SPECTRUM CHEMICAL MFG CORP
Warning

Synonyms: 2-(2-Butoxyethoxy)ethanol, Diethylene glycol mono-n-butyl ether, Butyldigol

B1178-200LTB B1178-1LTGL B1178-20LTMT
700003-458EA 4361.95 USD
700003-458 700003-456 700003-460
Diethylene glycol monobutyl ether ≥99%
Diethylene glycol monobutyl ether

2-(2-Butoxyethoxy)ethanol is more commonly known as Diethylene glycol butyl ether (DGBE) and is a type of glycol ether. It is primarily used as a solvent in coatings, inks, cleaners and specialty fluids.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: CH₃(CH₂)₃OCH₂CH₂OCH₂CH₂OH
MW: 162.23 g/mol
Boiling Pt: 230.4 °C (1013 hPa)
Melting Pt: –68 °C
Density: 0.954 g/cm³ (20 °C)
Flash Pt: 100 °C
Storage Temperature: Ambient
MDL Number: MFCD00002881
CAS Number: 112-34-5
EINECS: 203-961-6
Merck Index: 13,01556

Order Now

Specification Test Results

Assay 99% Min.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR