2-Butoxyethanol ≥97%
Supplier: SPECTRUM CHEMICAL MFG CORP
|
Warning
|
Synonyms:
Ethylene glycol butyl ether, Butyl cellosolve, Ethylene glycol monobutyl ether
2-Butoxyethanol is a colorless, sweet ether-like smelling liquid, which dissolves both water soluble and hydrophobic substances. It is a glycol ether with modest surfactant properties. A solvent of low toxicity, it is also relatively inexpensive and often used in household and industrial products. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.
Formula:
CH₃(CH₂)₃OCH₂CH₂OH MW: 118.18 g/mol Boiling Pt: 171 °C (1013 hPa) Melting Pt: –70 °C Density: 0.902 g/cm³ (20 °C) Flash Pt: 61 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002884 CAS Number: 111-76-2 EINECS: 203-905-0 UN: 2810 ADR: 6.1,III Merck Index: 13,01558 |
Specification Test Results
Assay | 97% Min. |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...