Benzotriazole ≥98%

Supplier: SPECTRUM CHEMICAL MFG CORP
Danger

Synonyms: 1,2,3-Benzotriazole, 1H-1,2,3-Benzotriazole, 1H-Benzotriazole, 1H-Benzo[d][1,2,3]triazole

B1089-100GM B1089-12KGBL B1089-500GM
700008-910EA 88.2 USD
700008-910 700008-912 700008-914
Benzotriazole ≥98%
Benzotriazole

Benzotriazole is a versatile compound that has been used as a restrainer in photographic emulsions, a reagent for the analytical determination of silver and has been extensively used as a corrosion inhibitor in the atmosphere and underwater.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: C₆H₅N₃
MW: 119.13 g/mol
Boiling Pt: 350 °C (1013 hPa)
Melting Pt: 99 °C
Density: 1.34 g/cm³ (20 °C)
Flash Pt: 170 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00005699
CAS Number: 95-14-7
EINECS: 202-394-1
UN: 2811
ADR: 6.1,III
Merck Index: 13,01109
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d0ba54cc42ef5ad9dbae6d7f118a3af1ae642287&vid=ce57b3f0d2583e32220d1e191d0202ad452e6475&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 98% Min.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR