Barium chloride dihydrate ≥99%, crystals ACS for synthesis
Supplier: SPECTRUM CHEMICAL MFG CORP
|
Danger
|
Synonyms:
Barium dichloride
B1025-12KG
B1025-125GM
B1025-500GM
B1025-2.5KG
700001-112EA
1185.07
USD
700001-112
700001-110
700001-116
700001-114
Barium chloride dihydrate ≥99%, crystals ACS for synthesis
Barium chloride dihydrate
Barium Chloride, Dihydrate, Crystal, Reagent, ACS is a very common water soluble salt of barium. It has many laboratory uses, such as testing for sulfate ions as well as being used in pyrotechnics for Barium chlorides distinctive bright green color. As an ACS grade reagent, Spectrum Chemical manufactured compound is used as the quality standard against which other substances are grade and has met the toughest regulatory standards for quality and pureness
Formula:
BaCl₂·2H₂O MW: 244.27 g/mol Boiling Pt: 1560 Melting Pt: 960 Density: 3.86 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00149154 CAS Number: 10326-27-9 EINECS: 233-788-1 UN: 1564 ADR: 6.1 III Merck Index: 14,00971 |
Specification Test Results
Assay | 99% Min. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...