L(+)-Arabinose ≥99%

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: Pectinose, Pectin Sugar

AR110-100GM AR110-1KG AR110-25GM AR110-5KGBL
700009-630EA 258.7 USD
700009-630 700009-632 700009-634 700009-636
L(+)-Arabinose ≥99%
L(+)-Arabinose

L-Arabinose is an aldopentose and is found in nature as a component of biopolymers like hemicellulose and pectin. It is also a monosaccharide that contains five carbon atoms. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.

Formula: C₅H₁₀O₅
MW: 150.13 g/mol
Boiling Pt: 331 °C (1013 hPa)
Melting Pt: 158 - 160 °C
Density: 1.585 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00067709
CAS Number: 5328-37-0
EINECS: 226-214-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1ad925ac21d4f12371621b1c44dbb6cda0a12da8&vid=6b9738c515009e5720388ea307490836c3b8afa8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 99% Min.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR