Aluminium chloride hexahydrate ≥95% USP suitable for use as excipient
Supplier: SPECTRUM CHEMICAL MFG CORP
|
Warning
|
Synonyms:
Aluminum chloride hexahydrate
AL225-100GM
AL225-500GM
AL225-50KGBL
AL225-2.5KG
AL225-12KG
95031-724EA
105.75
USD
95031-724
95031-728
700001-084
95031-726
700001-082
Aluminium chloride hexahydrate ≥95% USP suitable for use as excipient
Aluminium chloride hexahydrate
Aluminum Chloride, Hexahydrate, USP is used for its antistatic characteristics in some excipient applications.
All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Formula:
AlCl₃·6H₂O MW: 241.43 g/mol Melting Pt: 100 Density: 2.39 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00149134 CAS Number: 7784-13-6 EINECS: 231-208-1 UN: 3260 ADR: 8 III |
Specification Test Results
Assay | 95% Min. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...