Agar, granular NF

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: Agar Agar

AG105-2.5KG AG105-500GM
95031-648EA 1446.2 USD
95031-648 95031-650
Agar, granular NF
Agar

Form: Granular


All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP). 

Melting Pt: 90 °C
Storage Temperature: Ambient
MDL Number: MFCD00081288
CAS Number: 9002-18-0
EINECS: 232-658-1
Merck Index: 13,00184
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=36877cbcfaca6a0ace648ee20ae51b3467f2748c&vid=4295a0dcbc54bbbd374a5fb934297574c6848d32&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR