Adenine (synthetic) ≥99%

Supplier: SPECTRUM CHEMICAL MFG CORP
Danger

Synonyms: Purin-6-amine, 7H-Purin-6-amine, 6-Aminopurine

AD105-100GM AD105-25GM AD105-1KGBL AD105-5GM
700008-896EA 622.6 USD
700008-896 700008-900 700008-898 700008-902
Adenine (synthetic) ≥99%
Adenine

Adenine, Synthetic, also known as 6-aminopurine, is a nucleobase with many roles in biochemistry including cellular respiration. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.

Formula: C₅H₅N₅
MW: 135.13 g/mol
Density: 0.99741 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00041790
CAS Number: 73-24-5
EINECS: 200-796-1
UN: 2811
ADR: 6.1,III
Merck Index: 13,00152

Order Now

Specification Test Results

Assay 99% Min.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR