Adipohydrazide ≥99% for synthesis
Supplier: SPECTRUM CHEMICAL MFG CORP
Synonyms:
Adipyl hydrazide
A3802-25GM
A3802-100GM
700008-002EA
82.15
USD
700008-002
700008-000
Adipohydrazide ≥99% for synthesis
Adipohydrazide
Adipic Acid Dihydrazide, also known as ADH or Adipohydrazide, can be used as a hardener for epoxy resins and for cross-linking water-based emulsions. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.
Formula:
C₆H₁₄N₄O₂ MW: 174.2 g/mol Boiling Pt: 446 Melting Pt: 181…182 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007614 CAS Number: 1071-93-8 EINECS: 213-999-5 |
Specification Test Results
Assay | 99% Min. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...