AEROSIL® 200

Supplier: Spectrum Chemical Mfg. Corp.

A1801-500GM A1801-200KGB A1801-2.5KG
700000-682EA 273.16 USD
700000-682 700000-680 700000-678
AEROSIL® 200
AEROSIL®

Aerosil(R) 200, also known as fumed silica, is used as an anticaking agent in powders and a thickening agent in solutions.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Boiling Pt: 2230 °C (1013 hPa)
Melting Pt: 1719 °C
Density: 2.2 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011232
CAS Number: 112945-52-5
EINECS: 231-545-4
Merck Index: 14,08493

Order Now

Specification Test Results

specific surface area 200 m2/g


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR