Agar, powder FCC

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: Agar Agar

A1672-125GM A1672-500GM A1672-2.5KG
95031-656EA 326.4 USD
95031-656 95031-652 95031-654
Agar, powder FCC
Agar

Agar, Powder, FCC is a gel made from the cell walls of several species of red algae or seaweed. It appears as an odorless, tasteless, beige powder. It functions as a binder and thickener in cooking to make jellies, pudding, and custards. Spectrum Chemical offers over 300 Food Grade (FCC) chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities

Melting Pt: 90 °C
Storage Temperature: Ambient
MDL Number: MFCD00081288
CAS Number: 9002-18-0
EINECS: 232-658-1
Merck Index: 13,00184
https://pr.vwr.com/stibo/web/std.lang.all/81/91/2298191.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR