Amyl acetate ≥98%, Reagent Grade
Supplier: SPECTRUM CHEMICAL MFG CORP
|
Danger
|
Synonyms:
n-Pentyl acetate, Acetic acid amyl ester, Acetic acid pentyl ester, Pentyl acetate
A1280-500MLG
A1280-20LTMT
A1280-25ML
A1280-4LTGL
A1280-200LTB
700004-414EA
173.35
USD
700004-414
700004-408
700004-410
700004-412
700004-406
Amyl acetate ≥98%, Reagent Grade
Amyl acetate
Amyl Acetate, Reagent, with an odor similar to bananas and apples, is often used as a flavoring agent. It can also be used as a solvent and as part of the preparation of penicillin. Its reagent grade means this is the highest quality commercially available for this chemical and that the American Chemical Society has not officially set any specifications for this material.
Formula:
C₇H₁₄O₂ MW: 130.19 g/mol Boiling Pt: 149 °C (1013 hPa) Melting Pt: -71 °C Density: 0.876 g/cm³ (20 °C) Flash Pt: 23 °C Storage Temperature: Ambient |
MDL Number:
MFCD00009500 CAS Number: 628-63-7 EINECS: 211-047-3 UN: 1104 ADR: 3,III |
Specification Test Results
Assay | 98% Min. |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...