Phosphoryl trichloride ≥99.999% (P basis), PURATREM, Electronic Grade
Supplier: STREM CHEMICALS INC MS
|
Danger
|
97-8875-100G
97-8875-25G
100200-360EA
1017.66
USD
100200-360
100200-362
Phosphoryl trichloride ≥99.999% (P basis), PURATREM, Electronic Grade
Phosphoryl trichloride
Halides for CVD & ALD, Phosphine, Volatile Precursors for CVD
Formula:
POCl₃ Boiling Pt: 107 °C (1013 hPa) Melting Pt: 1 °C Density: 1.645 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011443 CAS Number: 10025-87-3 EINECS: 233-046-7 UN: 1810 ADR: 8,I Merck Index: 12,07506 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...