Germanium(IV) oxide ≥99.9999% (Ge basis), PURATREM, Electronic Grade
Supplier: STREM CHEMICALS INC MS
|
Warning
|
Synonyms:
Germanium dioxide
97-3222-50G
97-3222-10G
100202-476EA
711.94
USD
100202-476
100205-806
Germanium(IV) oxide ≥99.9999% (Ge basis), PURATREM, Electronic Grade
Germanium(IV) oxide
Formula:
GeO₂ MW: 104.61 g/mol Melting Pt: 1111…1119 °C Density: 3.64 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011030 CAS Number: 1310-53-8 EINECS: 215-180-8 Merck Index: 12,04417 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...