Boron tribromide ≥99.999% (B basis), PURATREM, Electronic Grade

Supplier: STREM CHEMICALS INC MS
Danger

97-1725-25G 97-1725-100G
100199-210EA 485.48 USD
100199-210 100207-200
Boron tribromide ≥99.999% (B basis), PURATREM, Electronic Grade
Boron tribromide

Halides for CVD & ALD, Volatile Precursors for CVD

Formula: BBr₃
MW: 250.52 g/mol
Boiling Pt: 91.2
Melting Pt: -46
Density: 2.6431
Storage Temperature: Ambient
MDL Number: MFCD00011312
CAS Number: 10294-33-4
EINECS: 233-657-9
UN: 2692
ADR: 8 I
Merck Index: 13,01337

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR