Antimony (V) oxide ≥99.998% (Sb basis), PURATREM, Electronic Grade
Supplier: STREM CHEMICALS INC MS
|
Danger
|
Synonyms:
Antimony pentoxide, Diantimony pentoxide
97-0382-100G
97-0382-25G
100200-356EA
533.6
USD
100200-356
100206-066
Antimony (V) oxide ≥99.998% (Sb basis), PURATREM, Electronic Grade
Antimony (V) oxide
Formula:
O₅Sb₂ Melting Pt: 2000 °C Density: 3.8 g/cm³ (20 °C) |
MDL Number:
MFCD00011216 CAS Number: 1314-60-9 EINECS: 215-237-7 UN: 1549 ADR: 6.1,III Merck Index: 12,00739 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...