Aluminium isopropoxide ≥99.99% (Al basis), PURATREM

Supplier: STREM CHEMICALS INC MS
Danger

Synonyms: Aluminiumtriisopropylate, Aluminiumisopropylate, Aluminiumtriisopropoxide, Aluminium isopropoxide

97-0139-25G 97-0139-100G
100210-596EA 253.35 USD
100210-596 100199-208
Aluminium isopropoxide ≥99.99% (Al basis), PURATREM
Aluminium isopropoxide

Alkoxide, Volatile Precursors for CVD

Formula: C₉H₂₁AlO₃
MW: 204.25 g/mol
Boiling Pt: 135
Melting Pt: 119
Density: 1.0346
MDL Number: MFCD00008870
CAS Number: 555-31-7
EINECS: 209-090-8
UN: 3178
ADR: 4.1 II
Merck Index: 13,00346

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR