Thorium tetrafluoride, anhydrous ≥99.9% (Th basis)

Supplier: STREM CHEMICALS INC MS
Danger

93-9002-50G 93-9002-10G
100202-340EA 379.32 USD
100202-340 100204-454
Thorium tetrafluoride, anhydrous ≥99.9% (Th basis)
Thorium tetrafluoride
Formula: F₄Th
MW: 308.03 g/mol
Boiling Pt: 1680 °C (1013 hPa)
Melting Pt: > 900 °C
Density: 6.1 g/cm³ (20 °C)
MDL Number: MFCD00036276
CAS Number: 13709-59-6
EINECS: 237-259-6
UN: 2910
ADR: 7,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR