Bismuth triacetate ≥99%
Supplier: STREM CHEMICALS INC MS
Synonyms:
Acetic acid bismuth (III) salt, Bismuth (III) acetate, Bismuth(III) acetate
93-8352-25G
93-8352-100G
100197-272EA
232.12
USD
100197-272
100208-606
Bismuth triacetate ≥99%
Bismuth triacetate
Formula:
(H₃CCOO)₃Bi MW: 386.11 g/mol |
MDL Number:
MFCD00015629 CAS Number: 22306-37-2 EINECS: 244-904-5 |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...