Bismuth(III) sulphide ≥99.999% (Bi basis), PURATREM
Supplier: STREM CHEMICALS INC MS
|
Warning
|
Synonyms:
Dibismuth trisulphide
93-8351-10G
93-8351-50G
100208-250EA
142.95
USD
100208-250
100203-878
Bismuth(III) sulphide ≥99.999% (Bi basis), PURATREM
Bismuth(III) sulphide
Formula:
Bi₂S₃ MW: 514.16 g/mol Melting Pt: 763 (dec.) Density: 7.39 |
MDL Number:
MFCD00014204 CAS Number: 1345-07-9 EINECS: 215-716-0 Merck Index: 12,01329 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...