Triphenylbismuth ≥99%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Bismuth triphenyle, Triphenylbismuthine

93-8350-50G 93-8350-10G
100210-590EA 621.35 USD
100210-590 100202-338
Triphenylbismuth ≥99%
Triphenylbismuth

Volatile Organometallics for CVD & ALD, Volatile Precursors for CVD

Formula: C₁₈H₁₅Bi
Boiling Pt: 208 °C (0.07 -rr)
Melting Pt: 78 °C
Density: 1.5851 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00014064
CAS Number: 603-33-8
EINECS: 210-033-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR