Bismuth ≥99.999%, powder
Supplier: STREM CHEMICALS INC MS
|
Warning
|
93-8348-50G
93-8348-10G
100203-606EA
356.68
USD
100203-606
100200-614
Bismuth ≥99.999%, powder
Bismuth
Formula:
Bi Boiling Pt: 1560 °C Melting Pt: 271.3 °C Density: 9.78 g/cm³ (20 °C) |
MDL Number:
MFCD00134033 CAS Number: 7440-69-9 EINECS: 231-177-4 UN: 3089 ADR: 4,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...