Bismuth ≥99.9%, powder
Supplier: STREM CHEMICALS INC MS
|
Warning
|
93-8347-250G
93-8347-50G
100205-802EA
165.6
USD
100205-802
100205-804
Bismuth ≥99.9%, powder
Bismuth
Formula:
Bi Boiling Pt: 1560 °C Melting Pt: 271.3 °C Density: 9.78 g/cm³ (20 °C) |
MDL Number:
MFCD00134033 CAS Number: 7440-69-9 EINECS: 231-177-4 UN: 3089 ADR: 4,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...