Bismuth ≥99.99%, shot, elongated

Supplier: STREM CHEMICALS INC MS

93-8346-500G 93-8346-100G
100198-474EA 366.58 USD
100198-474 100202-336
Bismuth ≥99.99%, shot, elongated
Bismuth
Formula: Bi
MW: 208.98 g/mol
Boiling Pt: 1560 °C
Melting Pt: 271.3 °C
Density: 9.80
MDL Number: MFCD00134033
CAS Number: 7440-69-9
EINECS: 231-177-4
Merck Index: 12,01297

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR