Bismuth(III) chloride, anhydrous ≥99.999% (Bi basis), PURATREM
Supplier: STREM CHEMICALS INC MS
|
Warning
|
Synonyms:
Bismuth(III) chloride, Bismuth trichloride
93-8315-5G
93-8315-25G
100209-234EA
92
USD
100209-234
100200-814
Bismuth(III) chloride, anhydrous ≥99.999% (Bi basis), PURATREM
Bismuth(III) chloride
Halides for CVD & ALD
Formula:
BiCl₃ MW: 315.34 g/mol Boiling Pt: 447 °C Melting Pt: 230…232 °C Density: 4.75 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00003461 CAS Number: 7787-60-2 EINECS: 232-123-2 UN: 3260 ADR: 8,III Merck Index: 13,01261 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...