Bismuth(III) chloride, anhydrous ≥99% (99.9+%-Bi)

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Bismuth(III) chloride, Bismuth trichloride

93-8314-25G 93-8314-100G
100211-308EA 103.32 USD
100211-308 100200-220
Bismuth(III) chloride, anhydrous ≥99% (99.9+%-Bi)
Bismuth(III) chloride

Halides for CVD & ALD

Formula: BiCl₃
MW: 315.34 g/mol
Boiling Pt: 447 °C
Melting Pt: 230…232 °C
Density: 4.75 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003461
CAS Number: 7787-60-2
EINECS: 232-123-2
UN: 3260
ADR: 8,III
Merck Index: 13,01261

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR