Bismuth(III) bromide ≥98%

Supplier: STREM CHEMICALS INC MS
Danger

93-8313-10G 93-8313-50G
100202-332EA 72.18 USD
100202-332 100206-064
Bismuth(III) bromide ≥98%
Bismuth(III) bromide
Formula: BiBr₃
MW: 448.69 g/mol
Boiling Pt: 453 °C
Melting Pt: 218 °C
Density: 5.72 g/cm³ (20 °C)
MDL Number: MFCD00014195
CAS Number: 7787-58-8
EINECS: 232-121-1
UN: 3260
ADR: 8,III
Merck Index: 12,01299

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR