Bismuth (III) sulfide ≥99.9% (Bi basis)
Supplier: STREM CHEMICALS INC MS
|
Warning
|
93-8309-100G
93-8309-25G
100208-246EA
700.62
USD
100208-246
100208-248
Bismuth (III) sulfide ≥99.9% (Bi basis)
Formula:
Bi₂S₃ Melting Pt: 685 °C (dec.) Density: 7.39 |
MDL Number:
MFCD00014204 CAS Number: 1345-07-9 EINECS: 215-716-0 Merck Index: 12,01329 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...